Order Entry
Norway
ContactUsLinkComponent
 
Analytical standard, Glufosinate-ammonium, reference material, Fluka™
  HONCR1707-100MG
 :  
Analytical standard, Glufosinate-ammonium, reference material, Fluka™
  HONCR1707-100MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    77182-82-2
  • Description:
    Glufosinate-ammonium
  • Description:
    Glufosinate-ammonium
  • Form:
    Neat