Nitrile gloves, Showa 7502PF
Supplier: Showa
Certificates
About this item
Designed for sensitive skin, the 7502PF nitrile disposable glove is powder-, latex- and accelerator-free.
- Biodegradable
- Ambidextrous
- 0,06 mm nitrile provides good chemical protection from a wide array of chemical hazards
- Large dispenser size: 180/200 gloves per dispenser
Featuring SHOWA’s revolutionary Eco Best Technology (EBT), these gloves are engineered for accelerated biodegradation in biologically active landfills.
Recommended for: Handling plants and vegetables, light chemical handling in horticulture, agriculture, food packing and handling, food service/food prep, washing and cleaning.
Certifications: EN ISO 374-1:2016 Type C (KT), EN 374-5 VIRUS; Cat. III
Food contact approved. GreenCircle certified. Biodegradation test results acc. to the norm ASTM D5526.
Food contact approved. GreenCircle certified. Biodegradation test results acc. to the norm ASTM D5526.
Packaging: 200 gloves per dispenser (size XXL: 180 gloves per dispenser). The dispenser is made of 85 % recycled cardboard, the outer carton is made of 90 % recycled material.
Specifications
- Catalog No:
- 112-6599
- 112-6598
- 112-6597
- 112-6596
- 112-6595
- 112-6594
- Model:
- 7502PF
- 7502PF
- 7502PF
- 7502PF
- 7502PF
- 7502PF
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Environmentally Preferable:
- Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
- Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
- Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
- Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
- Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
- Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- EN norm:
- EN ISO 374-1:2016 Type C (KT), EN 374-5 VIRUS
- EN ISO 374-1:2016 Type C (KT), EN 374-5 VIRUS
- EN ISO 374-1:2016 Type C (KT), EN 374-5 VIRUS
- EN ISO 374-1:2016 Type C (KT), EN 374-5 VIRUS
- EN ISO 374-1:2016 Type C (KT), EN 374-5 VIRUS
- EN ISO 374-1:2016 Type C (KT), EN 374-5 VIRUS
- Food contact approved:
- +
- +
- +
- +
- +
- +
- Environmental certification:
- Biodegradation test results acc. to the norm ASTM D5526. Green circle
- Biodegradation test results acc. to the norm ASTM D5526. Green circle
- Biodegradation test results acc. to the norm ASTM D5526. Green circle
- Biodegradation test results acc. to the norm ASTM D5526. Green circle
- Biodegradation test results acc. to the norm ASTM D5526. Green circle
- Biodegradation test results acc. to the norm ASTM D5526. Green circle