Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Aspergillus
niger
Pectinase
Aspergillus niger Pectinase
Aspergillus niger Pectinase
Supplier:
TCI
Certificates
Expand image
Aspergillus niger Pectinase
Supplier:
TCI
About this item
Specifications
Species:
Aspergillus niger
Enzyme name:
Pectinase
Specifications
Related Information
Documentation
Manual
Manual Bioscience and Analyticalscience
Datasheet
TCIAP0026-25G_TDS_EN
Documentation