Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Pig Lipase (from Pancreas)
Pig Lipase (from Pancreas)
Pig Lipase (from Pancreas)
Catalog # TCIAL0057-25G
Supplier:
TCI
CAS Number:
Certificates
Expand image
Pig Lipase (from Pancreas)
Catalog # TCIAL0057-25G
Supplier:
TCI
CAS Number:
Certificates
Specifications
Pk:
25 g
Source:
Pancreas
Species:
Pig
Enzyme name:
Lipase
Specifications
About this item
Product Details
Documentation
Product Details
Documentation