Order Entry
Norway
ContactUsLinkComponent
Bovine deoxyribonuclease (from Pancreas)
Bovine deoxyribonuclease (from Pancreas)
  TCIAD5564-100MG
 :  TCI
 :  
Bovine deoxyribonuclease (from Pancreas)
  TCIAD5564-100MG
 :  TCI
 :  

 

  • Pk:
    100 mg
  • Source:
    Pancreas
  • Species:
    Bovine
  • Enzyme name:
    Deoxyribonuclease