Order Entry
Norway
ContactUsLinkComponent
1-Cetylpyridinium bromide
1-Cetylpyridinium bromide
Katalog # MOLEM42597278
Supplier:  Molekula
CAS Number:  
1-Cetylpyridinium bromide
Katalog # MOLEM42597278
Supplier:  Molekula
CAS Number:  

Specifications

  • Pakketype:
    Glass bottle for solids
  • Fp:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :384.44 g/mol
Melting Pt:63…66 °C
EINECS:205-428-3
UN:2811
ADR:6.1,II
CAS Number:140-72-7
Formula:C₂₁H₃₈BrN
MDL Number:MFCD00011988