About this item
Innovative multi-layer chemical barrier technology, offering Type 3/4/5 protection.
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Designed to protect, typical innovative features include dual zip systems and double cuffs
- Welded and taped seams, our highest barrier to liquids and particulates
- Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
- Anti-static, tested according to EN 1149-5
Coveralls are designed to protect, with innovative features that include a 2-piece hood, double cuffs, and double zip closure. Elasticated hood, waist, and boot overflaps. Coveralls also feature integrated socks with boot overflaps and attached Ansell barrier gloves. Garments are manufactured from multi-layer chemical barrier material, with welded and taped seams.
Recommended for contaminated environments, chemical handling, filling/blending/charging raw materials, transferring liquids and solids, opening and draining pumps valves or lines, loading and unloading of proces equipment, raw material sample collection, concentration mixing and palletisation, Hazmat response, and large pipe assembly/disassembly.
Specifications
- Catalog No:
- ANSEGR40T-00125-08
- ANSEGR40T-00125-09
- ANSEGR40T-00125-04
- ANSEGR40T-00125-02
- ANSEGR40T-00125-03
- ANSEGR40T-00125-05
- ANSEGR40T-00125-06
- ANSEGR40T-00125-07
- Model:
- 125
- 125
- 125
- 125
- 125
- 125
- 125
- 125
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection
- Chemical protection