Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iron (III) acetylacetonate
Iron(III) acetylacetonate
Iron(III) acetylacetonate
:
Thermo Fisher Scientific
Iron(III) acetylacetonate
:
Thermo Fisher Scientific
:
14024-18-1
:
:
C₁₅H₂₁FeO₆
:
MFCD00000020
:
353.18 g/mol
:
100 °C (9.7513 mmHg)
:
∼ 183 °C (dec.)
:
5.24 g/cm³ (20 °C)
:
Ambient
:
237-853-5
:
GHS07
msds_ALFA012534
Thermo Scientific Acroseal 25 Years