Iron(II) iodide, anhydrous ≥97% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7783-86-0
:
Formula:FeI₂
MDL Number:MFCD00016094
:309.66 g/mol
Merck Index:12,04097
Density:5.32
Storage Temperature:Ambient
EINECS:232-031-2
UN:3260
ADR:8,III
:GHS05
:GHS07