Order Entry
Norway
ContactUsLinkComponent
1,5-Bis(diphenylphosphino)pentane
1,5-Bis(diphenylphosphino)pentane
Catalog # MOLE36583793-1G
Supplier:  Molekula
CAS Number:  
1,5-Bis(diphenylphosphino)pentane
Catalog # MOLE36583793-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :440.5 g/mol
Boiling Pt:230 °C (0.1 mmHg)
Boiling Pt:230 °C (0,1 mmHg)
Melting Pt:43…46 °C
CAS Number:27721-02-4
Formula:C₂₉H₃₀P₂
MDL Number:MFCD00003052