Arsenic(III) sulphide ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1303-33-9
:
Formula:As₂S₃
MDL Number:MFCD00003435
:246.04 g/mol
Merck Index:14,806
Boiling Pt:707 °C
Melting Pt:300 °C
Density:3.43
Storage Temperature:Ambient
EINECS:215-117-4
UN:1557
ADR:6.1,III
:GHS06
:GHS09