Calcium chloride ≥89%, pellets, purified, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:10043-52-4
:
Formula:CaCl₂
MDL Number:MFCD00010903
Merck Index:14,01659
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:772 °C
Density:2.15 g/cm³
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
REACH:01-2119494219-28
:GHS07
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:≥89%
- Assay (CaCl₂):≥89 %
- Heavy Metals (as Pb):≤0.005 %
- Iron (Fe):≤0.02 %
- Trace Impurities - Arsenic (As):≤1 ppm