
Propyl-3,4,5-trihydroxybenzoate ≥98% (by HPLC) for histology and pathology, for diagnostic, Sigma-Aldrich®
Catalog # 02370-100G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance (colour):White to faint brown
- Appearance (form):Powder
- Conforms to structure:≥98%
- undefined:Infrared spectrum
Specification Test Results
About this item
:
:212.2 g/mol
Merck Index:12,08044
Boiling Pt:449 °C (1013 hPa)
Melting Pt:145…148 °C
Density:1.21 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:204-498-2
:GHS07
CAS Number:121-79-9
Formula:C₁₀H₁₂O₅
MDL Number:MFCD00002196