Ethyl-3,4,5-trihydroxybenzoate for syntese, Sigma-Aldrich®
8.20597.0100
: Merck
:
- Pakketype:Plastic bottle
- Fp:100 g
- Seq:3401
- Solubility:(20 °C) slightly soluble
- Melting Point:150 - 152 °C
- Molar Mass:198.17 g/mol
:
:198.18 g/mol
:M-Clarity™ quality level = MQ200
:212-608-5
:831-61-8
:C₉H₁₀O₅
:MFCD00016430