Uridine 5′-monophosphate disodium salt ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:3387-36-8
:MFCD00149422
Formula:C₉H₁₁N₂Na₂O₉P
MDL Number:MFCD00006525
:368.15 g/mol
Storage Temperature:Refrigerator
EINECS:222-211-9
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:-11.0 to -17.0 deg (C=1, H2O) (calcd.on anh.substance)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together