Order Entry
Norway
ContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) solution for ELISA, ready-to-use
3,3',5,5'-Tetramethylbenzidine (TMB) solution for ELISA, ready-to-use
Catalog # TCIAT3854-100ML
Supplier:  TCI
CAS Number:  
3,3',5,5'-Tetramethylbenzidine (TMB) solution for ELISA, ready-to-use
Catalog # TCIAT3854-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorlessclear liquid
  • Peroxidase detection test:
    to pass test

Specification Test Results

About this item

 :
 :240.35 g/mol
Storage Temperature:Refrigerator
CAS Number:54827-17-7
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748