
Tributylhexylphosphonium bromide ≥98.0% (by titrimetric analysis)
: TCI
:105890-71-9
:
:C₁₈H₄₀BrP
:MFCD00134170
:367.40 g/mol
:GHS07
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %