1-Stearoyl-sn-glycero-3-phosphocholine ≥98.0% (by HPLC, titration analysis)
TCIAT3808-250MG
: TCI
:
- Pk:250 mg
- Seq:2590
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC) (CAD):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
:
:523.69 g/mol
:Freezer
:19420-57-6
:C₂₆H₅₄NO₇P
:MFCD00050427