3,4,5-Trifluorophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:827614-70-0
:
Formula:C₁₂H₁₄BF₃O₂
MDL Number:MFCD05663885
:258.0446 g/mol
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together