Order Entry
Norway
ContactUsLinkComponent
4,4',4''-Tris(diphenylamino)triphenylamine ≥97.0% (by HPLC, total nitrogen)
4,4',4''-Tris(diphenylamino)triphenylamine ≥97.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
undefined
4,4',4''-Tris(diphenylamino)triphenylamine ≥97.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    245.0 to 249.0 deg-C

Specification Test Results