
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:265651-18-1
:
Formula:C₉H₁₆F₆N₃O₃P
MDL Number:MFCD01863753
:359.21 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %