Order Entry
Norway
ContactUsLinkComponent
 
2,3,5-Trifluorophenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAT2874-5G
Supplier:  TCI
CAS Number:  
undefined
2,3,5-Trifluorophenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAT2874-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    97.0 to 112.0 %

Specification Test Results

About this item

 :
 :175.9 g/mol
 :GHS07
CAS Number:247564-73-4
Formula:C₆H₄BF₃O₂
MDL Number:MFCD01863166