
N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide ≥98.0% (by GC, total nitrogen)
Catalog # TCIAT2713-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:111.0 to 115.0 deg-C
Specification Test Results
About this item
:
:290.48 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:66065-85-8
Formula:C₇H₆Cl₃NO₅
MDL Number:MFCD00075216