Triamcinolone acetonide ≥98.0% (by HPLC and UV method)
Supplier: TCI
Certificates
About this item
CAS Number:76-25-5
:
Formula:C₂₄H₃₁FO₆
MDL Number:MFCD00056834
:434.504 g/mol
Merck Index:14,09596
Melting Pt:294°C
EINECS:200-948-7
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (UV-vis method):min. 98.0 % (after drying)
- Specific rotation [a]20/D:+100.0 to +107.0 deg (C=1, Dioxane)
- Solubility in Dioxane:almost transparency