
Tetradecyl acrylate ≥95.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:21643-42-5
:
Formula:C₁₇H₃₂O₂
MDL Number:MFCD00673244
:268.44 g/mol
Boiling Pt:138°C
Melting Pt:14°C
:Stabilised with 4-methoxyphenol (MEHQ)
EINECS:244-491-1
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %