Order Entry
Norway
ContactUsLinkComponent
Tri-tert-butylphosphonium tetraphenylborate ≥98.0% (by HPLC, titration analysis)
Toxic
Tri-tert-butylphosphonium tetraphenylborate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT2258-1G
Supplier:  TCI
CAS Number:  
Toxic
Tri-tert-butylphosphonium tetraphenylborate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT2258-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Iodometric Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :522.56 g/mol
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:131322-08-2
Formula:C₃₆H₄₈BP