
Thiomorpholinoacetic acid-1',1'-dioxide monohydrate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:155480-08-3
:
Formula:C₆H₁₁NO₄S
MDL Number:MFCD00121261
:193.22 g/mol
Melting Pt:177°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in hot Water:almost transparency
- Water:7.5 to 9.5 %