(m-Terphenyl-5'-yl)trimethylsilane ≥98.0% (by GC)
TCIAT1871-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:79.0 to 83.0 deg-C
:
:302.49 g/mol
:128388-53-4
:C₂₁H₂₂Si
:MFCD03844807