Order Entry
Norway
ContactUsLinkComponent
cis-3-Hexen-1-yltiglate ≥97.0% (by GC)
Irritant
cis-3-Hexen-1-yltiglate ≥97.0% (by GC)
Catalog # TCIAT0934-25ML
Supplier:  TCI
CAS Number:  
Irritant
cis-3-Hexen-1-yltiglate ≥97.0% (by GC)
Catalog # TCIAT0934-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :182.26 g/mol
Flash Pt:96°C
EINECS:267-554-5
 :GHS07
CAS Number:67883-79-8
Formula:C₁₁H₁₈O₂
MDL Number:MFCD00036528