

Testosterone Propionate ≥98.0% (by HPLC and UV method)
Supplier: TCI
Certificates
About this item
CAS Number:57-85-2
:
Formula:C₂₂H₃₂O₃
MDL Number:MFCD00003653
:344.50 g/mol
Merck Index:14,09180
EINECS:200-351-1
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (UV-vis method):min. 98.0 %
- Melting point:120.0 to 123.0 deg-C
- Specific rotation [a]20/D:+83.0 to +90.0 deg (C=1, Dioxane)