Calcium mesotartrate trihydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:815-79-2
:
Formula:C₄H₆O₆
MDL Number:MFCD00151483
:188.15 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 98.0 % (calcd.on anh.substance)
- Water:20.0 to 25.0 %