10H-Spiro[acridine-9,9'-fluorene] ≥98.0% (by HPLC, total nitrogen)
TCIAS0984-1G
: TCI
:
- Pk:1 g
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
:
:331.42 g/mol
:92638-81-8
:C₂₅H₁₇N
:MFCD29919320