Sulbutiamine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3286-46-2
:
Formula:C₃₂H₄₆N₈O₆S₂
MDL Number:MFCD01726427
:702.89 g/mol
Merck Index:14,09297
Storage Temperature:Refrigerator
EINECS:221-937-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %