

N-Succinimidyl 4-Azido-2,3,5,6-tetrafluorobenzoate ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:126695-58-7
:
Formula:C₁₁H₄F₄N₄O₄
MDL Number:MFCD00467437
:332.17 g/mol
UN:1325
ADR:4.1,III
:GHS02
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Melting point:110.0 to 114.0 deg-C