Order Entry
Norway
ContactUsLinkComponent
N-Succinimidyl 4-Formylbenzoate ≥98.0% (by HPLC)
N-Succinimidyl 4-Formylbenzoate ≥98.0% (by HPLC)
  TCIAS0893-100MG
 :  TCI
 :  
undefined
N-Succinimidyl 4-Formylbenzoate ≥98.0% (by HPLC)
  TCIAS0893-100MG
 :  TCI
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    168.0 to 175.0 deg-C

 

 

 :
 :247.21 g/mol
 :Freezer
 :60444-78-2
 :C₁₂H₉NO₅
 :MFCD00133733