
4-(Succinimidylcarboxy)benzophenone ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:91990-88-4
:
Formula:C₁₈H₁₃NO₄
MDL Number:MFCD00058572
:323.2995 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:207.0 to 211.0 deg-C