Order Entry
Norway
ContactUsLinkComponent
SATP (N-Succinimidyl 3-(Acetylthio)propionate) ≥95.0% (by GC)
SATP (N-Succinimidyl 3-(Acetylthio)propionate) ≥95.0% (by GC)
Catalog # TCIAS0859-100MG
Supplier:  TCI
CAS Number:  
undefined
SATP (N-Succinimidyl 3-(Acetylthio)propionate) ≥95.0% (by GC)
Catalog # TCIAS0859-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    64.0 to 69.0 deg-C

Specification Test Results

About this item

 :
 :245.25 g/mol
Storage Temperature:Refrigerator
CAS Number:84271-78-3
Formula:C₉H₁₁NO₅S
MDL Number:MFCD00133023