N-Succinimidyl Ferrocenecarboxylate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:115223-09-1
:
Formula:C₁₅H₁₃FeNO₄
MDL Number:MFCD03427488
:327.12 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Chelometric Titration):min. 97.0 %