Order Entry
Norway
ContactUsLinkComponent
Succinimidyl (2R)-6-(Tetrahydro-2H-pyran-2-yloxy)-2,5,7,8-tetramethylchroman-2-carboxylate
Succinimidyl (2R)-6-(Tetrahydro-2H-pyran-2-yloxy)-2,5,7,8-tetramethylchroman-2-carboxylate
Catalog # TCIAS0599-50MG
Supplier:  TCI
CAS Number:  
Succinimidyl (2R)-6-(Tetrahydro-2H-pyran-2-yloxy)-2,5,7,8-tetramethylchroman-2-carboxylate
Catalog # TCIAS0599-50MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Effect test of HPLC derivatization:
    to pass test

Specification Test Results

About this item

 :
 :431.49 g/mol
Storage Temperature:Refrigerator
CAS Number:1069137-73-0
Formula:C₂₃H₂₉NO₇