Order Entry
Norway
ContactUsLinkComponent
 
Irritant
Penicillin G potassium salt, Supelco®
Catalog # 46609-250MG
Supplier:  Merck
CAS Number:  
undefined
Irritant
Penicillin G potassium salt, Supelco®
Catalog # 46609-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :372.49 g/mol
Merck Index:12,07225
Melting Pt:214…217 °C
Density:1.359 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:204-038-0
 :GHS07
Storage Temperature:Refrigerator
CAS Number:113-98-4
Formula:C₁₆H₁₇KN₂O₄S
MDL Number:MFCD00036193