
Silybin (mixture of silybin A and silybin B) ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:36804-17-8
:
Formula:C₂₅H₂₂O₁₀
:482.44 g/mol
Merck Index:14,08532
Melting Pt:160°C
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Orange to Green powder to crystaline
- Purity (HPLC):min. 98.0 area% (total of isomers)
Specification Test Results
Frequently Bought Together