Order Entry
Norway
ContactUsLinkComponent
MBS (N-Succinimidyl 3-maleimidobenzoate) ≥98.0% (by HPLC, total nitrogen) cross-linking reagent
Irritant
MBS (N-Succinimidyl 3-maleimidobenzoate) ≥98.0% (by HPLC, total nitrogen) cross-linking reagent
Supplier:  TCI
Certificates
Irritant
MBS (N-Succinimidyl 3-maleimidobenzoate) ≥98.0% (by HPLC, total nitrogen) cross-linking reagent
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    178.0 to 183.0 deg-C

Specification Test Results

Related Information