Order Entry
Norway
ContactUsLinkComponent
 
Didodecyl succinate ≥93.0% (by GC)
Catalog # TCIAS0255-25G
Supplier:  TCI
CAS Number:  
Didodecyl succinate ≥93.0% (by GC)
Catalog # TCIAS0255-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 93.0 %
  • Melting point:
    38.0 to 43.0 deg-C

Specification Test Results

About this item

 :
 :454.74 g/mol
Storage Temperature:Refrigerator
EINECS:227-789-6
CAS Number:5980-15-4
Formula:C₂₈H₅₄O₄
MDL Number:MFCD00059284