Order Entry
Norway
ContactUsLinkComponent
Rapamycin ≥98.0% (by HPLC)
Health
Rapamycin ≥98.0% (by HPLC)
Catalog # TCIAR0097-25MG
Supplier:  TCI
CAS Number:  
Health
Rapamycin ≥98.0% (by HPLC)
Catalog # TCIAR0097-25MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    1.20 to 1.70 %
  • Specific rotation [a]20/D:
    -145 to -155 deg (C=0.2, MeOH)

Specification Test Results

About this item

 :
 :914.19 g/mol
Merck Index:14,08114
 :GHS08
Storage Temperature:Freezer
CAS Number:53123-88-9
Formula:C₅₁H₇₉NO₁₃
MDL Number:MFCD00867594