1-Pyrenebutanoic acid Succinimidyl ester ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:114932-60-4
:
Formula:C₂₄H₁₉NO₄
MDL Number:MFCD00077403
:385.42 g/mol
Melting Pt:133°C
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%