1-Palmitoyl-sn-glycero-3-phosphocholine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAP2694-250MG
Supplier: TCI
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC) (CAD):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+3.0 to +5.0 deg (C=0.1, methanol)
Specification Test Results
About this item
:
:495.638 g/mol
Melting Pt:253°C
Storage Temperature:Freezer
CAS Number:17364-16-8
Formula:C₂₄H₅₀NO₇P
MDL Number:MFCD00036904