

Procarbazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:366-70-1
:
Formula:C₁₂H₁₉N₃O·HCl
MDL Number:MFCD00072082
Merck Index:14,07758
Melting Pt:226°C
:257.76 g/mol
EINECS:206-678-6
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %