Order Entry
Norway
ContactUsLinkComponent
γ-(N-Morpholino)propanesulphonic acid (MOPS), Millipore®
γ-(N-Morpholino)propanesulphonic acid (MOPS), Millipore®
Catalog # 475922-100
CAS Number:  
γ-(N-Morpholino)propanesulphonic acid (MOPS), Millipore®
Catalog # 475922-100
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :209.27 g/mol
Merck Index:13,06288
Melting Pt:> 250 °C
Flash Pt:110 °C (closed cup)
Storage Temperature:Ambient
EINECS:214-478-5
CAS Number:1132-61-2
Formula:C₇H₁₅NO₄S
MDL Number:MFCD00006183