Order Entry
Norway
ContactUsLinkComponent
Phenyl-2,4,6-tri-O-acetyl-3-O-allyl-1-thio-β-D-glucopyranoside ≥98.0% (by HPLC)
Phenyl-2,4,6-tri-O-acetyl-3-O-allyl-1-thio-β-D-glucopyranoside ≥98.0% (by HPLC)
Catalog # TCIAP1736-1G
Supplier:  TCI
CAS Number:  
Phenyl-2,4,6-tri-O-acetyl-3-O-allyl-1-thio-β-D-glucopyranoside ≥98.0% (by HPLC)
Catalog # TCIAP1736-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    113.0 to 117.0 deg-C

Specification Test Results

About this item

 :
 :438.49 g/mol
Storage Temperature:Refrigerator
CAS Number:197005-22-4
Formula:C₂₁H₂₆O₈S