γ-(N-Morpholino)propanesulphonic acid (MOPS) 99% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1132-61-2
:
Formula:C₇H₁₅NO₄S
MDL Number:MFCD00006183
:209.27 g/mol
Merck Index:13,06288
Melting Pt:277…282 °C
Flash Pt:116 °C
Storage Temperature:Ambient
EINECS:214-478-5
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤1.0 %
- UV:(1 M in water) 1 cm cell
- pH:3.0 to 4.5 (1 % in water at 25°C)
- Titration with NaOH:≥98.5 %